Learn More
Waters Corp Waters Vial, LCGC Certified, Clear Glass, 15 × 45 mm, Screw Neck, 3 mL Total Recovery
Catalog No. 50782554 Shop All Waters Corporation Products
Encompass_Preferred
Certified vials are tested for cleanliness by HPLC and held to the tightest dimensional tolerances in the industry.
  • Total Recovery vial format
  • Screw neck closure
  • Designed for sample preparation
  • Made from clear glass
  • Dimensions of 15 × 45 mm
  • 3 mL volume capacity
  • Preslit PTFE/silicone seal material
  • Black cap
  • Autosampler vial type
  • LCGC certified
  • Includes vial, cap, & septum
  • Sample limited
Catalog No. 50-782-554 Supplier Waters Corp Supplier No. 186002630C
Certified vials are tested for cleanliness by HPLC and held to the tightest dimensional tolerances in the industry.
  • Total Recovery vial format
  • Screw neck closure
  • Designed for sample preparation
  • Made from clear glass
  • Dimensions of 15 × 45 mm
  • 3 mL volume capacity
  • Preslit PTFE/silicone seal material
  • Black cap
  • Autosampler vial type
  • LCGC certified
  • Includes vial, cap, & septum
  • Sample limited