Learn More
Chemglass Life Sciences 3mL Micro Volumetric Flask, Class A, Tolerance: +-0.02mL
SDP
Catalog No. CG160303
Encompass_Preferred
3mL Micro Volumetric Flask, Class A, Tolerance: ¿0.02mL. Class ¿A¿ micro volumetric flask calibrated to contain. With 14/20 top outer joint and supplied complete with a CG-3000 14/20 hollow stopper. Flasks have hex base for stability and ¿V¿ shaped interior bottom for total sample retrieval.
Catalog No. CG160303 Supplier Chemglass Life Sciences Supplier No. CG160303
3mL Micro Volumetric Flask, Class A, Tolerance: ¿0.02mL. Class ¿A¿ micro volumetric flask calibrated to contain. With 14/20 top outer joint and supplied complete with a CG-3000 14/20 hollow stopper. Flasks have hex base for stability and ¿V¿ shaped interior bottom for total sample retrieval.