Learn More
Bellco Glass, Inc. 3ml Pasteur Pipette, Individually Wrapped, sterile 500/pk, 2000/cs
SDP
Supplier:  Bellco Glass, Inc. 451400302
Encompass_Preferred
3ml Pasteur Pipette, Individually Wrapped, sterile 500/pk, 2000/cs. Converntional consumables for quantitative transfer of liquids, in the field of experimental direction of pipetting operations such as biology and chemistry. Cell culture laboratory liquid tranfer, dosing, aspiration or other liquid transfer. • Material: medical grade transparent LDPE• Excellent transparency with clear graduations for easy observation• Good fluidity for easy control• High precision and drop repeatability• Sterilized by Ethylene Oxide
Catalog No. 50-232-9750