Learn More
Chemglass Life Sciences Pasteur Pipette, 3mL, Bulk Packed, NON-Sterile
SDP
Catalog No. 501897720
Encompass_Preferred
Material: Medical grade transparent LDPEExcellent transparency with clear graduations for easy observationGood fluidity for easy controlHigh precision and drop repeatabilityBulk PackedDNase/RNase, endotoxin and pyrogenic free
Catalog No. 50-189-7720 Supplier Chemglass Life Sciences Supplier No. CGN3404003
Material: Medical grade transparent LDPEExcellent transparency with clear graduations for easy observationGood fluidity for easy controlHigh precision and drop repeatabilityBulk PackedDNase/RNase, endotoxin and pyrogenic free