Learn More
Bellco Glass, Inc. 3mL Pipette, MLA, Macro, Green
SDP
Catalog No. 502329731
Encompass_Preferred
3mL Pipette, MLA, Macro, GreenSingle-stroke “to deliver” operationAvailable in 10 fixed volumes ranging from 1 mL- 10 mLAlso available as 1-5 mL Adjustable model with 0.1 mL incrementsAlso available as Selectable model for set volumes: 2 mL/3 mL/4 mL/5 mL All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorGreenPrimary MaterialAluminum Alloy & Stainless SteelVolume3mLVistaLab Tip Fit SizeMacro 5Universal FitNoAccuracy±0.75%Precision0.50%
Catalog No. 50-232-9731 Supplier Bellco Glass, Inc. Supplier No. 451900082
3mL Pipette, MLA, Macro, GreenSingle-stroke “to deliver” operationAvailable in 10 fixed volumes ranging from 1 mL- 10 mLAlso available as 1-5 mL Adjustable model with 0.1 mL incrementsAlso available as Selectable model for set volumes: 2 mL/3 mL/4 mL/5 mL All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorGreenPrimary MaterialAluminum Alloy & Stainless SteelVolume3mLVistaLab Tip Fit SizeMacro 5Universal FitNoAccuracy±0.75%Precision0.50%