Learn More
Cole-Parmer BD Biocoat Disposable Syringe, Non-Sterile, Slip-Tip, Bulk Pack, 3 mL, 1600/Cs
Catalog No. NC1490058 Shop All Cole-Parmer Products
Encompass
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-Lok or slip-tip versions. Syringes come nonsterilized in bulk packages and can be autoclaved. Needles and shield tips are not included; order separately.
Catalog No. NC1490058 Supplier Cole-Parmer Supplier No. 0794504
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-Lok or slip-tip versions. Syringes come nonsterilized in bulk packages and can be autoclaved. Needles and shield tips are not included; order separately.