Learn More
CELLTREAT Scientific Products 3mL Transfer Pipet, Individually Wrapped, Sterile
SDP
Catalog No. 5000144312
Encompass_Preferred
Effortlessly transfer solutions without the need for any additional equipmentPolyethylene Transfer Pipets are easy to manipulate with a gentle squeezeTranslucent and flexible pipet body is ideal for a range of liquid handling applicationsEach pipet features molded graduations with labeled incrementsSterile Transfer Pipets are conveniently individually wrapped in plastic and are suitable for sterile solutions or cell suspensions
Catalog No. 50-001-44312 Supplier CELLTREAT Scientific Products Supplier No. 229214
Effortlessly transfer solutions without the need for any additional equipmentPolyethylene Transfer Pipets are easy to manipulate with a gentle squeezeTranslucent and flexible pipet body is ideal for a range of liquid handling applicationsEach pipet features molded graduations with labeled incrementsSterile Transfer Pipets are conveniently individually wrapped in plastic and are suitable for sterile solutions or cell suspensions