Learn More
Chemglass Life Sciences Chemglass 3 mL Sterile Transport Tubes with Separated Caps, 200/Bag (CGN-4742-23)
SDP
Catalog No. 502733399
Encompass_Preferred
Chemglass 3 mL transport tubes with separated caps are sterile, RNase- and pyrogen-free sample containers designed for safe storage, transport, and freezing of biological and environmental samples. Electron-beam sterilized to SAL 10⁻⁶ and manufactured in a 100,000-level clean workshop, they are suitable for clinical, environmental, food testing, and research laboratory workflows.
  • Sterile, electron-beam sterilized to SAL 10⁻⁶.
  • RNase-free, DNase-free, pyrogen-free, and endotoxin-free.
  • USP VI polypropylene tubes with HDPE caps.
  • Separated caps for flexible sample handling.
  • Packaging: 200 tubes per bag and 200 caps per bag; 1,000 tubes and 1,000 caps per case.
  • Temperature range suitable from -196°C to 121°C.
  • Excellent sealing performance; rated against 95 kPa negative pressure to prevent leakage during transport.
  • Produced in a 100,000-level clean workshop for controlled manufacturing conditions.
Catalog No. 50-273-3399 Supplier Chemglass Life Sciences Supplier No. CGN474223
Add to Cart
Add to Cart
Chemglass 3 mL transport tubes with separated caps are sterile, RNase- and pyrogen-free sample containers designed for safe storage, transport, and freezing of biological and environmental samples. Electron-beam sterilized to SAL 10⁻⁶ and manufactured in a 100,000-level clean workshop, they are suitable for clinical, environmental, food testing, and research laboratory workflows.
  • Sterile, electron-beam sterilized to SAL 10⁻⁶.
  • RNase-free, DNase-free, pyrogen-free, and endotoxin-free.
  • USP VI polypropylene tubes with HDPE caps.
  • Separated caps for flexible sample handling.
  • Packaging: 200 tubes per bag and 200 caps per bag; 1,000 tubes and 1,000 caps per case.
  • Temperature range suitable from -196°C to 121°C.
  • Excellent sealing performance; rated against 95 kPa negative pressure to prevent leakage during transport.
  • Produced in a 100,000-level clean workshop for controlled manufacturing conditions.