Learn More
S2 Media 3mL vLogiX ™ (VSTM), 16x115mm Polypropylene Tube with Swab Capture, 50/box
Catalog No. 502995890
Encompass_Preferred
vLogiX ™ (VTSM) is a non-selective and non-differential culture medium intended for the collection, conveyance, and storage of clinical specimens containing viral agents. vLogiX ™ (VTSM) is intended for use in standard laboratory procedures for culture and diagnostic assays.• Product meets ASM, CDC, and CLSI reference methods and performance criteria.• vLogiX™ should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5890 Supplier S2 Media Supplier No. 5165
vLogiX ™ (VTSM) is a non-selective and non-differential culture medium intended for the collection, conveyance, and storage of clinical specimens containing viral agents. vLogiX ™ (VTSM) is intended for use in standard laboratory procedures for culture and diagnostic assays.• Product meets ASM, CDC, and CLSI reference methods and performance criteria.• vLogiX™ should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.