Learn More
Ace Glass, Inc. 3ML VOLUMETRIC BULB 3ML VOLUMETRIC BULB
SDP
Catalog No. 50979611
Encompass_Preferred
3mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.
Catalog No. 50-979-611 Supplier Ace Glass, Inc. Supplier No. 800421
3mL volumetric bulb for use with ACE Automatic Dispensing Pipet. Bulbs have a short tipping angle and give reproduceable volumes.