Learn More
Sutter Instruments Box Filaments 3.0mm square box filament, 3.0mm wide
SDP
Catalog No. NC9452114
Encompass_Preferred
Box filaments are particularly suitable for patch pipettes, slice preparations, and for sharp electrodes and microinjection pipettes where gradual parallel walls would aid cell penetration. When using a box filament, the size of the square box should be approximately 1.0mm to 1.5mm larger than the outside diameter of the glass that you will be using.
Catalog No. NC9452114 Supplier Sutter Instruments Supplier No. FGFB330B
Box filaments are particularly suitable for patch pipettes, slice preparations, and for sharp electrodes and microinjection pipettes where gradual parallel walls would aid cell penetration. When using a box filament, the size of the square box should be approximately 1.0mm to 1.5mm larger than the outside diameter of the glass that you will be using.