Learn More
Electron Microscopy Sciences Flat Embedding Mold 3 mm with Cocoon Box
SDP
Catalog No. 50930534
Encompass_Preferred
No more stickiness with new storage boxes. Most resins are no longer a challenge to flat embed when used in combination with the Cocoon box. The system withstands temperatures up to 70 Degrees C, and our flat embedding of many resins is made easier.
Catalog No. 50-930-534 Supplier Electron Microscopy Sciences Supplier No. 70902CB
No more stickiness with new storage boxes. Most resins are no longer a challenge to flat embed when used in combination with the Cocoon box. The system withstands temperatures up to 70 Degrees C, and our flat embedding of many resins is made easier.