Learn More
Andwin Scientific 3MM GLASS BEADS 1LB/PKG
SDP
Catalog No. NC9775989
Encompass_Preferred
Solid Soda Lime Glass Beads, 3mm dia. no. 3000 glass beads. Chemical resistant.Specific gravity 2.49g/cm3, temperature of transformation, Tg440degreeC. Softening temperature Tt505degreeC, coefficient of linear extension 20 to 100degree
This item is not returnable. View return policy
Catalog No. NC9775989 Supplier Andwin Scientific Supplier No. 26396508
Solid Soda Lime Glass Beads, 3mm dia. no. 3000 glass beads. Chemical resistant.Specific gravity 2.49g/cm3, temperature of transformation, Tg440degreeC. Softening temperature Tt505degreeC, coefficient of linear extension 20 to 100degree