Learn More
Medipoint 3mm Goldenrod Animal Lancet
Catalog No. NC1826831
Encompass_Preferred
The 3mm point length is for use on rats. It is used only for pulling 1 or 2 drops of blood.
Catalog No. NC1826831 Supplier Medipoint Supplier No. MSPPGR3MM
The 3mm point length is for use on rats. It is used only for pulling 1 or 2 drops of blood.