Learn More
Chemglass Life Sciences Stopcock, Straight Bore, PTFE Plug, 3mm Bore, 15.2/30 Plug Size, 10mm OD Sidearms
SDP
Catalog No. 501531733
Encompass_Preferred
Stopcock, Straight Bore, PTFE Plug, 3mm Bore, 15.2/30 Plug Size, 10mm OD Sidearms. Straight bore stopcock having a 1:5 taper solid PTFE plug.
Catalog No. 50-153-1733 Supplier Chemglass Life Sciences Supplier No. CG42404
Stopcock, Straight Bore, PTFE Plug, 3mm Bore, 15.2/30 Plug Size, 10mm OD Sidearms. Straight bore stopcock having a 1:5 taper solid PTFE plug.