Learn More
Azzota Corp 10mm Pathlength Micro Cuvettes 1Ml - Pathlength 10mm - Inside Width 3mm Volume 1.0Ml Dimensions 45X12.5X12.5mm; Cover And Black Walls, Quartz (190Nm-2500Nm)
Catalog No. 507532906
Encompass_Preferred
10mm Pathlength Micro Cuvettes 1ml - Pathlength 10mm - Inside width 3mm Volume 1.0ml Dimensions 45x12.5x12.5mm; Cover and black walls, Quartz (190nm-2500nm)
Catalog No. 50-753-2906 Supplier Azzota Corp Supplier No. Q134
10mm Pathlength Micro Cuvettes 1ml - Pathlength 10mm - Inside width 3mm Volume 1.0ml Dimensions 45x12.5x12.5mm; Cover and black walls, Quartz (190nm-2500nm)