Learn More
Ace Glass, Inc. 3MM STOPCOCK GB 3MM STOPCOCK GB
SDP
Catalog No. 50979693
Encompass_Preferred
Glass 3mm straight bore stopcock with a glass plug and a tapered flange, which insures maximum strength at the seal. The bore of the side arms is equal to or slightly larger than the bore of the plug.
Catalog No. 50-979-693 Supplier Ace Glass, Inc. Supplier No. 813706
Glass 3mm straight bore stopcock with a glass plug and a tapered flange, which insures maximum strength at the seal. The bore of the side arms is equal to or slightly larger than the bore of the plug.