Learn More
Roboz Surgical Micro Dissecting Trephine, Tungsten Carbide, 2.7mm Diameter
SDP
Catalog No. NC9542057
Encompass
Micro Dissecting Trephine. Tungsten Carbide, 2.7mm Diameter. The head diameter of these trephines is 2.7 mm. The shaft diameter is 2.33 mm. They work with our Ideal micr odrill or other handpieces or drills accepting drill bits with 2.33 mm shaft dia
Catalog No. NC9542057 Supplier Roboz Surgical Supplier No. RS6310
Micro Dissecting Trephine. Tungsten Carbide, 2.7mm Diameter. The head diameter of these trephines is 2.7 mm. The shaft diameter is 2.33 mm. They work with our Ideal micr odrill or other handpieces or drills accepting drill bits with 2.33 mm shaft dia