Learn More
Cayman Chemical 3-Mercaptopicolinic Acid (hydrochloride), Purity: ≥95%, CAS: 320386-54-7, 100 mg
SDP
Catalog No. 502983204
Encompass_Preferred
An inhibitor of gluconeogenesis that targets PEPCK
Catalog No. 50-298-3204 Supplier Cayman Chemical Supplier No. 20895100 MG
An inhibitor of gluconeogenesis that targets PEPCK