Learn More
Cayman Chemical 3-methyl-α-Pyrrolidinobutiophenone (hydrochloride), Purity: ≥98%, 10 mg
SDP
Catalog No. 502892891
Encompass_Preferred
An isomer of 4-methyl-α-PBP, having the methyl group attached at the 3 position of the phenyl ring, instead of the 4 position; intended for forensic and research applications
Catalog No. 50-289-2891 Supplier Cayman Chemical Supplier No. 900118910 MG
An isomer of 4-methyl-α-PBP, having the methyl group attached at the 3 position of the phenyl ring, instead of the 4 position; intended for forensic and research applications