Learn More
Cayman Chemical 3-methyl-α-Pyrrolidinopropiophenone (hydrochloride), Purity: ≥98%, CAS: 2749302-38-1, 50 mg
SDP
Catalog No. 502951093
Encompass_Preferred
A structural isomer of methyl-α-PPP, having the methyl group at the 3 position, rather than the 4 position, of the phenyl group; intended for forensic and research applications
Catalog No. 50-295-1093 Supplier Cayman Chemical Supplier No. 1148550 MG
A structural isomer of methyl-α-PPP, having the methyl group at the 3 position, rather than the 4 position, of the phenyl group; intended for forensic and research applications