Learn More
Regis Technologies Inc 3N HCl in n-Butanol, 500 mL, colorless to light yellow liquid, alkylation reagent for GC derivatization
SDP
Catalog No. NC2868814
Encompass
3N HCl in n-Butanol, 500 mL, colorless to light yellow liquid, alkylation reagent for GC derivatization
Catalog No. NC2868814 Supplier Regis Technologies Inc Supplier No. 1201010200
3N HCl in n-Butanol, 500 mL, colorless to light yellow liquid, alkylation reagent for GC derivatization