Learn More
Andwin Scientific 3N - SODIUM HYDROXIDE 1L
SDP
Catalog No. NC0293551
Encompass_Preferred
Sodium Hydroxide solution has a concentration of 3.0 M. This compound is often used as a base, for chemical pulping, for tissue digestion, for dissolving amphoteric metals and compounds, as a esterification and transesterification reagent, and for food preparation as salt. This is packaged and sold in a 1 liter poly bottle.
Catalog No. NC0293551 Supplier Andwin Scientific Supplier No. BDH34721
Add to Cart
Add to Cart
Sodium Hydroxide solution has a concentration of 3.0 M. This compound is often used as a base, for chemical pulping, for tissue digestion, for dissolving amphoteric metals and compounds, as a esterification and transesterification reagent, and for food preparation as salt. This is packaged and sold in a 1 liter poly bottle.