Learn More
Cayman Chemical 3OAcetyl2deoxyadenosIn 2.7mg
SDP
Catalog No. 502919000
Encompass_Preferred
An acetylated derivative of dATP; has been used as a DNA synthesis chain termination reagent in Sanger sequencing
Catalog No. 50-291-9000 Supplier Cayman Chemical Supplier No. 385322.7 MG
Add to Cart
Add to Cart
An acetylated derivative of dATP; has been used as a DNA synthesis chain termination reagent in Sanger sequencing