Learn More
AdipoGen 3OH-C10-L-HSL
SDP
Catalog No. 502560749
Encompass_Preferred
Chemical. CAS 192883-12-8. Formula C14H25NO4. MW 271.35. Synthetic. N-3-hydroxydecanoyl-L-homoserine lactone is a small diffusible signaling molecule secreted by various bacteria and involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. This lactone is produced via lactonolysis from 3-oxodecanoyl-homoserine lactone, altering quorum sensing or contributing to quorum quenching. The applications of this molecule include regulation of virulence and exoproteases.
Catalog No. 50-256-0749 Supplier AdipoGen Supplier No. CDXH0086M025
Chemical. CAS 192883-12-8. Formula C14H25NO4. MW 271.35. Synthetic. N-3-hydroxydecanoyl-L-homoserine lactone is a small diffusible signaling molecule secreted by various bacteria and involved in quorum sensing, thereby controlling gene expression and affecting cellular metabolism. This lactone is produced via lactonolysis from 3-oxodecanoyl-homoserine lactone, altering quorum sensing or contributing to quorum quenching. The applications of this molecule include regulation of virulence and exoproteases.