Learn More
AdipoGen 3OH-C4-L-HSL
SDP
Supplier:  AdipoGen CDXH0084M025
Encompass_Preferred
Chemical. CAS 1325550-06-8. Formula C8H13NO4. MW 187.19. Synthetic 3-Hydroxy-butanoyl-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Catalog No. 50-256-0752