Learn More
Cayman Chemical 3'-O-Methylguanosine-5'-O-triphosphate (sodium salt), Purity: ≥95%, CAS: 98923-57-0, 2.7 mg
SDP
Catalog No. 502916351
Encompass_Preferred
A methylated derivative of GTP; has been used as a chain termination reagent in the preparation of early RNA polymerase II elongation intermediates
Catalog No. 50-291-6351 Supplier Cayman Chemical Supplier No. 386882.7 MG
A methylated derivative of GTP; has been used as a chain termination reagent in the preparation of early RNA polymerase II elongation intermediates