Learn More
Cayman Chemical 3OnthrnIloyl 2deoxyad 600ug
SDP
Catalog No. 502917878
Encompass_Preferred
A fluorescent nucleotide derivative; inhibits B. pertussis and B. anthracis ACs (Kis = 9.1 and 8.0 µM, respectively); has been used as a probe for binding sites in, as well as ion effects on the enzymatic activity of, B. pertussis and B. anthracis ACs; ex/em = 330/425 nm, respectively
Catalog No. 50-291-7878 Supplier Cayman Chemical Supplier No. 38442600 UG
A fluorescent nucleotide derivative; inhibits B. pertussis and B. anthracis ACs (Kis = 9.1 and 8.0 µM, respectively); has been used as a probe for binding sites in, as well as ion effects on the enzymatic activity of, B. pertussis and B. anthracis ACs; ex/em = 330/425 nm, respectively