Learn More
Cayman Chemical 3OnthrnIloyl2deoxygua 600ug
SDP
Catalog No. 502918945
Encompass_Preferred
A fluorescent nucleotide derivative; has been used to probe inhibitor and activator binding sites on UMP kinase isolated from B. subtilis; ex/em = 330/450 nm, respectively
Catalog No. 50-291-8945 Supplier Cayman Chemical Supplier No. 38547600 UG
A fluorescent nucleotide derivative; has been used to probe inhibitor and activator binding sites on UMP kinase isolated from B. subtilis; ex/em = 330/450 nm, respectively