Learn More
Uline Cold Packs - 3 oz,1 EA
Catalog No. NC9896036
Encompass
Reusable leakproof refrigerant gel stays colder than ice. Freeze before use. Dependable, durable and reusable. Thick 4 mil poly. FDA compliant. Solid when frozen, not flexible. Gel is non-toxic, food safe. Can be used to prevent freezing if heated in a microwave. Heat on low setting for 30 second increments. Flip pack over every 30 seconds until warm. Outer bag can be recycled. Cut open to remove the gel. Gel can be disposed of in garbage. Do not place down the drain. Do not consume. Do not apply to the body. Ideal placement is on top of product. Not instant cold packs. Place in freezer up to 72 hours before they are frozen and ready to use. Cannot be frozen in ultra-low freezer. Poly exterior can withstand temperatures of 30 F.
Catalog No. NC9896036 Supplier Uline Supplier No. S13376
Reusable leakproof refrigerant gel stays colder than ice. Freeze before use. Dependable, durable and reusable. Thick 4 mil poly. FDA compliant. Solid when frozen, not flexible. Gel is non-toxic, food safe. Can be used to prevent freezing if heated in a microwave. Heat on low setting for 30 second increments. Flip pack over every 30 seconds until warm. Outer bag can be recycled. Cut open to remove the gel. Gel can be disposed of in garbage. Do not place down the drain. Do not consume. Do not apply to the body. Ideal placement is on top of product. Not instant cold packs. Place in freezer up to 72 hours before they are frozen and ready to use. Cannot be frozen in ultra-low freezer. Poly exterior can withstand temperatures of 30 F.