Learn More
MSC Leviton 2711 125/250 VAC 30A NEMA L14-30P Industrial Twist Lock Plug 3 Poles, 4 Wires, 1 Phase, Self-Grounding, 0.595 to 1.15" Cord Diam, Black/White
Catalog No. 501298393
Encompass
Leviton 2711 125/250 VAC 30A NEMA L14-30P Industrial Twist Lock Plug 3 Poles, 4 Wires, 1 Phase, Self-Grounding, 0.595 to 1.15" Cord Diam, Black/White
Catalog No. 50-129-8393 Supplier MSC Supplier No. 67585265
Leviton 2711 125/250 VAC 30A NEMA L14-30P Industrial Twist Lock Plug 3 Poles, 4 Wires, 1 Phase, Self-Grounding, 0.595 to 1.15" Cord Diam, Black/White