Learn More
Invivogen 3P-HPRNA
SDP
Catalog No. NC1434723
Encompass
5 triphosphate hairpin RNA; RIG-I ligand25ug Working Concentration: 10 ng- 1 g/ml
Catalog No. NC1434723 Supplier Invivogen Supplier No. TLRLHPRNA
5 triphosphate hairpin RNA; RIG-I ligand25ug Working Concentration: 10 ng- 1 g/ml