Learn More
MSC Delfin Vacuum, DG 75 AF, 11.5 HP, 3-Phase, 1 ea
Catalog No. 502816828
Encompass
The DG 75 AF is a high-performance, heavy-duty, three-phase industrial vacuum cleaner designed for continuous use in demanding industrial environments. It handles dust and solids, including combustible dust, featuring a maintenance-free side channel blower motor for high airflow. Constructed with powder-coated steel, it's compact and portable with a 26-gallon roll-out container for easy emptying.
  • Maintenance-free side channel blower motor for continuous operation
  • Removable 26-gallon collection tank with safety release
  • Large 53 ft² antistatic polyester star filter, class M
  • Manual shaker system for filter cleaning
  • Pressure relief valve for motor protection
  • Compact, portable design with powder-coated steel construction
  • Optional HEPA H14 filter available
Catalog No. 50-281-6828 Supplier MSC Supplier No. 13677174
Add to Cart
Add to Cart
Request a Quote
The DG 75 AF is a high-performance, heavy-duty, three-phase industrial vacuum cleaner designed for continuous use in demanding industrial environments. It handles dust and solids, including combustible dust, featuring a maintenance-free side channel blower motor for high airflow. Constructed with powder-coated steel, it's compact and portable with a 26-gallon roll-out container for easy emptying.
  • Maintenance-free side channel blower motor for continuous operation
  • Removable 26-gallon collection tank with safety release
  • Large 53 ft² antistatic polyester star filter, class M
  • Manual shaker system for filter cleaning
  • Pressure relief valve for motor protection
  • Compact, portable design with powder-coated steel construction
  • Optional HEPA H14 filter available