Learn More
MSC Delfin Industrial DG 50 SE, 3-Phase, 26 gal, 1 ea
Catalog No. 502817829
Encompass
The DG 50 SE is a highly performing industrial vacuum designed for both wet and dry applications. It is a compact yet powerful heavy-duty option, featuring a 2-stage side channel blower with two impellers in series, generating a high vacuum level suitable for moving heavier materials. Designed for continuous use, it requires no maintenance.
  • Compact and portable design
  • Constructed with powder-coated steel chassis
  • Side channel blower for continuous, maintenance-free operation
  • Removable 26-gallon collection tank with two-handed safety release
  • Large surface star filter with manual shaking system, Class L efficiency
  • Performance indicator light and pressure relief valve
  • Integrated cleaning system
  • Suitable for continuous heavy-duty extraction of dust, solids, and debris
Catalog No. 50-281-7829 Supplier MSC Supplier No. 13676036
Add to Cart
Add to Cart
Request a Quote
The DG 50 SE is a highly performing industrial vacuum designed for both wet and dry applications. It is a compact yet powerful heavy-duty option, featuring a 2-stage side channel blower with two impellers in series, generating a high vacuum level suitable for moving heavier materials. Designed for continuous use, it requires no maintenance.
  • Compact and portable design
  • Constructed with powder-coated steel chassis
  • Side channel blower for continuous, maintenance-free operation
  • Removable 26-gallon collection tank with two-handed safety release
  • Large surface star filter with manual shaking system, Class L efficiency
  • Performance indicator light and pressure relief valve
  • Integrated cleaning system
  • Suitable for continuous heavy-duty extraction of dust, solids, and debris