Learn More
MSC Delfin Industrial Vacuum, DG 50 SE, 6.4 HP, 26 gal
Catalog No. 502818016
Encompass
The DG 50 SE is a high-performance industrial vacuum designed for both wet and dry applications. It features a 2-stage side channel blower for high vacuum, suitable for continuous use, and includes a Class L filter with a manual shaking system for easy cleaning. Material is collected in a 26-gallon roll-out tank for convenient disposal.
  • Compact and portable design
  • Powder-coated steel construction
  • Maintenance-free side channel blower for continuous use
  • Removable, roll-out collection tank for easy disposal
  • Large surface Class L polyester star filter
  • Integrated manual filter shaking system
  • Pressure relief valve for motor protection
  • ABS accessory basket for storage
Catalog No. 50-281-8016 Supplier MSC Supplier No. 13676705
Add to Cart
Add to Cart
Request a Quote
The DG 50 SE is a high-performance industrial vacuum designed for both wet and dry applications. It features a 2-stage side channel blower for high vacuum, suitable for continuous use, and includes a Class L filter with a manual shaking system for easy cleaning. Material is collected in a 26-gallon roll-out tank for convenient disposal.
  • Compact and portable design
  • Powder-coated steel construction
  • Maintenance-free side channel blower for continuous use
  • Removable, roll-out collection tank for easy disposal
  • Large surface Class L polyester star filter
  • Integrated manual filter shaking system
  • Pressure relief valve for motor protection
  • ABS accessory basket for storage