Learn More
Echelon Biosciences Research Labs 3PHOSPHOTHIONATE PI(3)P 100ug
SDP
Catalog No. 50720607
Encompass
A synthetic phosphatidylinositol (3)-phospate [PI(3)P] analog in which the 3-phosphate has been rendered metabolically stable as a phosphothionate. P-3S18 has been shown to bind to the FYVE domain and is a substrate for the 5-kinase, PIKfyve. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-607 Supplier Echelon Biosciences Research Labs Supplier No. P3S18100UG
A synthetic phosphatidylinositol (3)-phospate [PI(3)P] analog in which the 3-phosphate has been rendered metabolically stable as a phosphothionate. P-3S18 has been shown to bind to the FYVE domain and is a substrate for the 5-kinase, PIKfyve. This item is non returnable