Learn More
Oxyrase Oxyrase OxyPRAS Plus 3-Pack, Selective Isolation, PRAS Media, 10 Multi-Packs
SDP
Catalog No. NC0519445
Encompass
The Oxyrase OxyPRAS Plus 3-Pack is a pre-reduced anaerobically sterilized culture media solution designed for selective isolation and cultivation of anaerobic microorganisms. It includes Brucella Blood Agar, PEA Blood Agar, and KVL Agar plates, ensuring enhanced recovery of injured cells. This product provides reliable results with a 3-month refrigerated shelf-life and 2-hour benchtop stability, making it essential for clinical anaerobes applications.
Catalog No. NC0519445 Supplier Oxyrase Supplier No. MBKP3
The Oxyrase OxyPRAS Plus 3-Pack is a pre-reduced anaerobically sterilized culture media solution designed for selective isolation and cultivation of anaerobic microorganisms. It includes Brucella Blood Agar, PEA Blood Agar, and KVL Agar plates, ensuring enhanced recovery of injured cells. This product provides reliable results with a 3-month refrigerated shelf-life and 2-hour benchtop stability, making it essential for clinical anaerobes applications.