Learn More
USA Scientific Inc 3-place angled pipette stand
Catalog No. 502745169
Encompass
White acrylic stand for 3 pipettes, including 1 multichannel position. 6" W x 10.5" high x 5" deep.
Catalog No. 50-274-5169 Supplier USA Scientific Inc Supplier No. 19990003
White acrylic stand for 3 pipettes, including 1 multichannel position. 6" W x 10.5" high x 5" deep.