Learn More
USA Scientific Inc 3-place upright freezer rack
Catalog No. 502745082
Encompass
3-place (3 long, no shelves) upright freezer rack for 5 1/4 in x 5 1/4 in boxes, 16 5/16 in L x 5 1/2 in W x 8 1/4 in H, stainless steel
Catalog No. 50-274-5082 Supplier USA Scientific Inc Supplier No. 26003230
3-place (3 long, no shelves) upright freezer rack for 5 1/4 in x 5 1/4 in boxes, 16 5/16 in L x 5 1/2 in W x 8 1/4 in H, stainless steel