Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3PO | 18550-98-6 | 99.9% | 210.23 | 1 ML
SDP

Catalog No. 5000400777
Encompass_Preferred

3PO is a light yellow to yellow solid and an inhibitor of PFKFB3. It attenuates the proliferation of several cancer cell lines with IC50s of 1.4-24 μmol/L. 3PO suppresses glucose uptake and decreases the intracellular concentration of Fru-2,6-BP, lactate, ATP, NAD+ and NADH. It can be used for the research of cancer.

  • Solid appearance
  • Purity of 99.85%
  • Inhibitor of PFKFB3
  • Attenuates proliferation of cancer cell lines
  • Suppresses glucose uptake
  • Decreases intracellular concentrations of Fru-2,6-BP, lactate, ATP, NAD+, and NADH
  • Soluble in DMSO at 60 mg/mL

Catalog No. 50-004-00777 Supplier Medchemexpress LLC Supplier No. HY1982410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3PO is a light yellow to yellow solid and an inhibitor of PFKFB3. It attenuates the proliferation of several cancer cell lines with IC50s of 1.4-24 μmol/L. 3PO suppresses glucose uptake and decreases the intracellular concentration of Fru-2,6-BP, lactate, ATP, NAD+ and NADH. It can be used for the research of cancer.

  • Solid appearance
  • Purity of 99.85%
  • Inhibitor of PFKFB3
  • Attenuates proliferation of cancer cell lines
  • Suppresses glucose uptake
  • Decreases intracellular concentrations of Fru-2,6-BP, lactate, ATP, NAD+, and NADH
  • Soluble in DMSO at 60 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.