Learn More
Medchemexpress LLC 3PO | 18550-98-6 | 99.9% | 210.23 | 50 MG
SDP
Catalog No. 5000401677
Encompass_Preferred
3PO is an inhibitor of PFKFB3 that has been shown to attenuate the proliferation of several cancer cell lines with IC50s ranging from 1.4 to 24 μmol/L. It suppresses glucose uptake and decreases the intracellular concentration of Fru-2,6-BP, lactate, ATP, NAD+, and NADH. This product is intended for cancer research.
  • Inhibits Jurkat cells proliferation by influencing cell cycle.
  • Suppresses glycolytic flux to lactate in Jurkat cells.
  • Inhibits cellular proliferation of transformed tumor cell lines.
  • Suppresses the growth of Lewis lung carcinoma xenografts in C57Bl/6 mice.
  • Suppresses xenograft tumorigenic growth of MDA-MB231 cells.
  • Suppresses xenograft tumorigenic growth of HL-60 leukemia.
  • Decreases Fru-2,6-BP and glucose uptake in C57Bl/6 mice.
Catalog No. 50-004-01677 Supplier Medchemexpress LLC Supplier No. HY1982450MG
3PO is an inhibitor of PFKFB3 that has been shown to attenuate the proliferation of several cancer cell lines with IC50s ranging from 1.4 to 24 μmol/L. It suppresses glucose uptake and decreases the intracellular concentration of Fru-2,6-BP, lactate, ATP, NAD+, and NADH. This product is intended for cancer research.
  • Inhibits Jurkat cells proliferation by influencing cell cycle.
  • Suppresses glycolytic flux to lactate in Jurkat cells.
  • Inhibits cellular proliferation of transformed tumor cell lines.
  • Suppresses the growth of Lewis lung carcinoma xenografts in C57Bl/6 mice.
  • Suppresses xenograft tumorigenic growth of MDA-MB231 cells.
  • Suppresses xenograft tumorigenic growth of HL-60 leukemia.
  • Decreases Fru-2,6-BP and glucose uptake in C57Bl/6 mice.