Learn More
TARGETMOL CHEMICALS INC 3PO 25MG
SDP
Catalog No. 502577197
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 3PO is a small-molecule inhibitor of PFKFB3 (IC50 22.9 uM) inhibiting the proliferation of several human malignant hematopoietic and adenocarcinoma cell lines (IC50 1.4-24 uM). It suppresses glucose uptake and decreases the intracellular concentration of Fru-26-BP lactate ATP NAD+ and NADH. purity: 99%
Catalog No. 50-257-7197 Supplier TARGETMOL CHEMICALS INC Supplier No. T2222325MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 3PO is a small-molecule inhibitor of PFKFB3 (IC50 22.9 uM) inhibiting the proliferation of several human malignant hematopoietic and adenocarcinoma cell lines (IC50 1.4-24 uM). It suppresses glucose uptake and decreases the intracellular concentration of Fru-26-BP lactate ATP NAD+ and NADH. purity: 99%