Learn More
Chemglass Life Sciences 3-Port Sparger Assy with Holes for 8L VSA Vessels
SDP
Catalog No. 50143737
Encompass
Our 3-port sparger assemblies with drilled holes, 0. 040” spaced 1/2” apart, and hose barbs are designed for vigorous aeration invertical side arm (VSA) vessels.These assemblies are constructed of 316L stainless steel, electro-polished to a 20Ra finish or better and include a blue polypropylene GL-45 open top cap and white silicone gasket.
Catalog No. 50-143-737 Supplier Chemglass Life Sciences Supplier No. CLS1437H42
Our 3-port sparger assemblies with drilled holes, 0. 040” spaced 1/2” apart, and hose barbs are designed for vigorous aeration invertical side arm (VSA) vessels.These assemblies are constructed of 316L stainless steel, electro-polished to a 20Ra finish or better and include a blue polypropylene GL-45 open top cap and white silicone gasket.