Learn More
Chemglass Life Sciences 3-Port Sparger Assy with Frit for 3L VSA Vessels
SDP
Catalog No. 50143789
Encompass
Our 3-port sparger assemblies are designed for gentle aeration using a 10 micron frit invertical side arm (VSA) vessels.The frit is removable for easy cleaning.These assemblies are constructed of 316L stainless steel, electro-polished to a 20Ra finish or better and include a blue polypropylene GL-45 open top cap and white silicone gasket.
Catalog No. 50-143-789 Supplier Chemglass Life Sciences Supplier No. CLS1437F50
Our 3-port sparger assemblies are designed for gentle aeration using a 10 micron frit invertical side arm (VSA) vessels.The frit is removable for easy cleaning.These assemblies are constructed of 316L stainless steel, electro-polished to a 20Ra finish or better and include a blue polypropylene GL-45 open top cap and white silicone gasket.