Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine 98.0+%, TCI America™
SDP

Catalog No. B29205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29205G 5 g
B29201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29205G Supplier TCI America Supplier No. B29205G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 147081-49-0
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD03427030
InChI Key CMIBWIAICVBURI-SSDOTTSWSA-N
Synonym r-+-1-boc-3-aminopyrrolidine, r-1-boc-3-aminopyrrolidine, r-3-amino-1-n-boc-pyrrolidine, r-tert-butyl 3-aminopyrrolidine-1-carboxylate, tert-butyl 3r-3-aminopyrrolidine-1-carboxylate, r-bocap, r-+-n-boc-3-aminopyrrolidine, 3r-3-amino-1-tert-butoxycarbonyl pyrrolidine, r-n-boc-3-aminopyrrolidine, r-3-amino-n-boc-pyrrolidine
PubChem CID 854070
IUPAC Name tert-butyl (3R)-3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)N

Specifications

Color Yellow
Boiling Point 244°C
Quantity 5 g
Formula Weight 186.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.