Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R)-(-)-3-Aminopyrrolidine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. A10535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10535G 5 g
A105325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10535G Supplier TCI America Supplier No. A10535G

Chemical Identifiers

CAS 116183-81-4
Molecular Formula C4H12Cl2N2
Molecular Weight (g/mol) 159.054
MDL Number MFCD00070604
InChI Key NJPNCMOUEXEGBL-RZFWHQLPSA-N
Synonym 3r---3-aminopyrrolidine dihydrochloride, r---3-aminopyrrolidine dihydrochloride, r-pyrrolidin-3-amine dihydrochloride, r-3-aminopyrrolidine 2hcl, r-+-3-aminopyrrolidine 2hcl, 3r-pyrrolidin-3-amine dihydrochloride, 3r-3-pyrrolidinamine dihydrochloride, r-3-aminopyrrolidine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, r, pubchem5730
PubChem CID 13500643
IUPAC Name (3R)-pyrrolidin-3-amine;dihydrochloride
SMILES C1CNCC1N.Cl.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 159.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (3R)-(-)-3-Aminopyrrolidine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.