Learn More
[(3R)-3-Hydroxyhexadecanoyl]-L-carnitine, ≥95% (TLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 1469901-03-8 |
| Linear Formula | C23H45 NO5 |
| Synonym | (2 R)-3-Carboxy-2-[[(3 R)-3-hydroxy-1-oxohexadecyl]oxy]-N,N,N-trimethyl-1-propanaminium inner salt |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 415.61 |
| Percent Purity | ≥95% (TLC) |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.