Learn More
Medchemexpress LLC (3R,4R)-4-(benzyloxy)-3-((tert-butoxycarbonyl)amino)pentanoic acid | 254101-11-6 | MFCD01862935 | ≥98.0% | 323.38 g/mol | C17H25NO5 | 250 MG
SDP
Catalog No. 5000283619
Encompass_Preferred
(3R,4R)-4-(benzyloxy)-3-((tert-butoxycarbonyl)amino)pentanoic acid is a threonine derivative supplied for laboratory research use. It is provided as a stable solid for in vitro experiments and should be handled using standard laboratory safety procedures; not for human or clinical use.
  • Threonine derivative useful for synthetic and biochemical research.
  • Stable solid form with recommended cold storage for long-term retention.
  • Molecular formula C17H25NO5 and molecular weight 323.38 g/mol.
  • Commonly available in small research-scale quantities for assay development.
  • Intended for research use only; follow appropriate handling and disposal procedures.
Catalog No. 50-002-83619 Supplier Medchemexpress LLC Supplier No. HYW140881250MG
(3R,4R)-4-(benzyloxy)-3-((tert-butoxycarbonyl)amino)pentanoic acid is a threonine derivative supplied for laboratory research use. It is provided as a stable solid for in vitro experiments and should be handled using standard laboratory safety procedures; not for human or clinical use.
  • Threonine derivative useful for synthetic and biochemical research.
  • Stable solid form with recommended cold storage for long-term retention.
  • Molecular formula C17H25NO5 and molecular weight 323.38 g/mol.
  • Commonly available in small research-scale quantities for assay development.
  • Intended for research use only; follow appropriate handling and disposal procedures.