Learn More
Medchemexpress LLC HY-111532 5mg Medchemexpress, (3R,4R)-A2-32-01 CAS:1359752-95-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1115325MG
Encompass_Preferred
Medchemexpress, HY-111532 5mg (3R,4R)-A2-32-01 CAS:1359752-95-6 (3R,4R)-A2-32-01 (compound 2), an anti-virulence drug, is a specific caseinolytic protein proteases (ClpP) inhibitor with an EC50 of 4.5 μM, and shows a tolerable cytotoxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9697