Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R,4S)-3-Hydroxy-4-phenyl-2-azetidinone 98.0+%, TCI America™
SDP

Catalog No. H1656250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1656250MG 250 mg
H16561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1656250MG Supplier TCI America Supplier No. H1656250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132127-34-5
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD07368374
InChI Key FBZSDKXFQUKDLD-JGVFFNPUSA-N
PubChem CID 10219589
IUPAC Name (3R,4S)-3-hydroxy-4-phenylazetidin-2-one
SMILES C1=CC=C(C=C1)C2C(C(=O)N2)O

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 250 mg
Formula Weight 163.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (3R,4S)-3-Hydroxy-4-phenyl-2-azetidinone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.